missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethoxynaphthalene 95.0+%, TCI America™
$126.76 - $384.00
Chemical Identifiers
| CAS | 5328-01-8 |
|---|---|
| Molecular Formula | C12H12O |
| Molecular Weight (g/mol) | 172.23 |
| MDL Number | MFCD00003928 |
| InChI Key | APWZAIZNWQFZBK-UHFFFAOYSA-N |
| Synonym | Ethyl 1-Naphthyl Ether |
| PubChem CID | 21403 |
| IUPAC Name | 1-ethoxynaphthalene |
| SMILES | CCOC1=CC=CC2=CC=CC=C21 |
Chemical Identifiers
| 5328-01-8 | |
| 172.23 | |
| APWZAIZNWQFZBK-UHFFFAOYSA-N | |
| 21403 | |
| CCOC1=CC=CC2=CC=CC=C21 |
| C12H12O | |
| MFCD00003928 | |
| Ethyl 1-Naphthyl Ether | |
| 1-ethoxynaphthalene |
Specifications
| 5328-01-8 | |
| MFCD00003928 | |
| Ethyl 1-Naphthyl Ether | |
| CCOC1=CC=CC2=CC=CC=C21 | |
| 172.23 | |
| 172.23 | |
| Liquid |
| C12H12O | |
| 5 g | |
| APWZAIZNWQFZBK-UHFFFAOYSA-N | |
| 1-ethoxynaphthalene | |
| 21403 | |
| ≥95.0% (GC) | |
| 1-Ethoxynaphthalene |
Safety and Handling
TSCA : No