missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethoxynaphthalene 95.0+%, TCI America™
Supplier: TCI America E053225G
Specifications
| 1-Ethoxynaphthalene | |
| C12H12O | |
| 25 g | |
| APWZAIZNWQFZBK-UHFFFAOYSA-N | |
| 1-ethoxynaphthalene | |
| 21403 | |
| ≥95.0% (GC) |
| 5328-01-8 | |
| MFCD00003928 | |
| Ethyl 1-Naphthyl Ether | |
| CCOC1=CC=CC2=CC=CC=C21 | |
| 172.23 | |
| 172.23 | |
| Liquid |
Chemical Identifiers
| 5328-01-8 | |
| 172.23 | |
| APWZAIZNWQFZBK-UHFFFAOYSA-N | |
| 21403 | |
| CCOC1=CC=CC2=CC=CC=C21 |
| C12H12O | |
| MFCD00003928 | |
| Ethyl 1-Naphthyl Ether | |
| 1-ethoxynaphthalene |
Safety and Handling
TSCA : No