missing translation for 'onlineSavingsMsg'
Learn More

(1-Ethoxycarbonylethyl)triphenylphosphonium bromide, 97%

CAS: 30018-16-7 | C23H24BrO2P | 443.32 g/mol

Supplier:  Thermo Scientific Chemicals A1302314

 View more versions of this product

Catalog No. AAA1302314


Only null left
Explore available promotions

Description

Description

1-Ethoxycarbonylethyl)triphenylphosphonium bromide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is widely used as an organophosphine catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethoxycarbonylethyl)triphenylphosphonium bromide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is widely used as an organophosphine catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Moisture sensitive. Incompatible to strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
Specifications

Specifications

(1-Ethoxycarbonylethyl)triphenylphosphonium bromide
∼130°C (decomposition)
MFCD00031605
3822244
1-ethoxy-1-oxopropan-2-yl triphenylphosphonium bromide, 1-ethoxycarbonyl ethyltriphenylphosphonium bromide, 1-ethoxycarbonyl ethyl triphenylphosphonium bromide, 2-ethoxy-1-methyl-2-oxoethyl triphenylphosphonium bromide, ceetppb, 1-carbethoxyethyl triphenylphosphonium bromide, 1-ethoxycarbonylethyl triphenylphosphonium bromide, 1-ethoxy-1-oxopropan-2-yl triphenylphosphanium bromide, phosphonium, 2-ethoxy-1-methyl-2-oxoethyl triphenyl-, bromide, ethoxycarbonylethyltriphenylphosphoniumbromide
RSYXORMKBUFAMS-UHFFFAOYNA-M
(1-ethoxy-1-oxopropan-2-yl)-triphenylphosphanium;bromide
3084511
97%
30018-16-7
C23H24BrO2P
25 g
Hygroscopic
Soluble in water.
[Br-].CCOC(=O)C(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
443.32
443.33

Chemical Identifiers

30018-16-7
443.32
RSYXORMKBUFAMS-UHFFFAOYNA-M
3084511
[Br-].CCOC(=O)C(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C23H24BrO2P
MFCD00031605
1-ethoxy-1-oxopropan-2-yl triphenylphosphonium bromide, 1-ethoxycarbonyl ethyltriphenylphosphonium bromide, 1-ethoxycarbonyl ethyl triphenylphosphonium bromide, 2-ethoxy-1-methyl-2-oxoethyl triphenylphosphonium bromide, ceetppb, 1-carbethoxyethyl triphenylphosphonium bromide, 1-ethoxycarbonylethyl triphenylphosphonium bromide, 1-ethoxy-1-oxopropan-2-yl triphenylphosphanium bromide, phosphonium, 2-ethoxy-1-methyl-2-oxoethyl triphenyl-, bromide, ethoxycarbonylethyltriphenylphosphoniumbromide
(1-ethoxy-1-oxopropan-2-yl)-triphenylphosphanium;bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 250-002-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only