missing translation for 'onlineSavingsMsg'
Learn More

(1-Ethoxycarbonylethyl)triphenylphosphonium bromide, 97%

Catalog No. p-7020910
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1302314 25 g
AAA1302322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1302314 Supplier Thermo Scientific Chemicals Supplier No. A1302314
Only null left

CAS: 30018-16-7 | C23H24BrO2P | 443.32 g/mol

1-Ethoxycarbonylethyl)triphenylphosphonium bromide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is widely used as an organophosphine catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethoxycarbonylethyl)triphenylphosphonium bromide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is widely used as an organophosphine catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Moisture sensitive. Incompatible to strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.

Chemical Identifiers

CAS 30018-16-7
Molecular Formula C23H24BrO2P
Molecular Weight (g/mol) 443.32
MDL Number MFCD00031605
InChI Key RSYXORMKBUFAMS-UHFFFAOYNA-M
Synonym 1-ethoxy-1-oxopropan-2-yl triphenylphosphonium bromide, 1-ethoxycarbonyl ethyltriphenylphosphonium bromide, 1-ethoxycarbonyl ethyl triphenylphosphonium bromide, 2-ethoxy-1-methyl-2-oxoethyl triphenylphosphonium bromide, ceetppb, 1-carbethoxyethyl triphenylphosphonium bromide, 1-ethoxycarbonylethyl triphenylphosphonium bromide, 1-ethoxy-1-oxopropan-2-yl triphenylphosphanium bromide, phosphonium, 2-ethoxy-1-methyl-2-oxoethyl triphenyl-, bromide, ethoxycarbonylethyltriphenylphosphoniumbromide
PubChem CID 3084511
IUPAC Name (1-ethoxy-1-oxopropan-2-yl)-triphenylphosphanium;bromide
SMILES [Br-].CCOC(=O)C(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼130°C (decomposition)
Quantity 25 g
Beilstein 3822244
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 443.33
Percent Purity 97%
Chemical Name or Material (1-Ethoxycarbonylethyl)triphenylphosphonium bromide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 250-002-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only