Learn More
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate, 98%
CAS: 52186-89-7 | C24H24BF4O2P | 462.23 g/mol
$402.49 - $1604.66
Chemical Identifiers
| CAS | 52186-89-7 |
|---|---|
| Molecular Formula | C24H24BF4O2P |
| Molecular Weight (g/mol) | 462.23 |
| MDL Number | MFCD00051879 |
| InChI Key | RGJYRMUGSAFITK-UHFFFAOYSA-N |
| Synonym | 1-carbethoxycyclopropyltriphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphanium tetrafluoroborate, acmc-20aowe, 1-carboethoxycyclopropyl triphenylphosphonium tetrafluoroborate, 1-carbethoxycyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphanium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphonium tetrafluoroborate |
| PubChem CID | 11016017 |
| SMILES | F[B-](F)(F)F.CCOC(=O)C1(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1830506
|
Thermo Scientific Chemicals
A1830506 |
5 g |
Each for $402.49
|
|
|||||
|
AAA1830514
|
Thermo Scientific Chemicals
A1830514 |
25 g |
Each for $1,604.66
|
|
|||||
Description
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 52186-89-7 | |
| 462.23 | |
| RGJYRMUGSAFITK-UHFFFAOYSA-N | |
| 11016017 |
| C24H24BF4O2P | |
| MFCD00051879 | |
| 1-carbethoxycyclopropyltriphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphanium tetrafluoroborate, acmc-20aowe, 1-carboethoxycyclopropyl triphenylphosphonium tetrafluoroborate, 1-carbethoxycyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphanium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphonium tetrafluoroborate | |
| F[B-](F)(F)F.CCOC(=O)C1(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
| 52186-89-7 | |
| C24H24BF4O2P | |
| 5 g | |
| 1-carbethoxycyclopropyltriphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphanium tetrafluoroborate, acmc-20aowe, 1-carboethoxycyclopropyl triphenylphosphonium tetrafluoroborate, 1-carbethoxycyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphanium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphonium tetrafluoroborate | |
| RGJYRMUGSAFITK-UHFFFAOYSA-N | |
| [1-(ethoxycarbonyl)cyclopropyl]triphenylphosphanium; tetrafluoroboranuide | |
| 11016017 | |
| 98% |
| 173°C to 179°C | |
| MFCD00051879 | |
| 4650673 | |
| Slightly soluble in water. | |
| F[B-](F)(F)F.CCOC(=O)C1(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 462.23 | |
| 462.23 | |
| (1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only