accessibility menu, dialog, popup

UserName

CAS RN 52186-89-7

IUPAC Name: [1-(ethoxycarbonyl)cyclopropyl]triphenylphosphanium; tetrafluoroboranuide
Synonyms: [1-(ethoxycarbonyl)cyclopropyl]triphenylphosphanium tetrafluoroborate
Molecular Weight (g/mol): 462.23
Molecular Formula: C24H24BF4O2P
InChi Key: RGJYRMUGSAFITK-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCOC(=O)C1(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results