missing translation for 'onlineSavingsMsg'
Learn More

(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate, 98%

Catalog No. AAA1830506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1830506 5 g
AAA1830514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1830506 Supplier Thermo Scientific Chemicals Supplier No. A1830506
Only null left

CAS: 52186-89-7 | C24H24BF4O2P | 462.23 g/mol

(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 52186-89-7
Molecular Formula C24H24BF4O2P
Molecular Weight (g/mol) 462.23
MDL Number MFCD00051879
InChI Key RGJYRMUGSAFITK-UHFFFAOYSA-N
Synonym 1-carbethoxycyclopropyltriphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphanium tetrafluoroborate, acmc-20aowe, 1-carboethoxycyclopropyl triphenylphosphonium tetrafluoroborate, 1-carbethoxycyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphanium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphonium tetrafluoroborate
PubChem CID 11016017
SMILES F[B-](F)(F)F.CCOC(=O)C1(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 173°C to 179°C
Quantity 5 g
Beilstein 4650673
Solubility Information Slightly soluble in water.
IUPAC Name [1-(ethoxycarbonyl)cyclopropyl]triphenylphosphanium; tetrafluoroboranuide
Formula Weight 462.23
Percent Purity 98%
Chemical Name or Material (1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only