missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Docosanol 98.0+%, TCI America™
$79.74 - $400.61
Chemical Identifiers
| CAS | 661-19-8 |
|---|---|
| Molecular Formula | C22H46O |
| Molecular Weight (g/mol) | 326.609 |
| MDL Number | MFCD00002939 |
| InChI Key | NOPFSRXAKWQILS-UHFFFAOYSA-N |
| Synonym | 1-docosanol, behenyl alcohol, docosanol, behenic alcohol, docosyl alcohol, n-docosanol, abreva, tadenan, lidavol, lanette 22 |
| PubChem CID | 12620 |
| ChEBI | CHEBI:31000 |
| IUPAC Name | docosan-1-ol |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCO |
Chemical Identifiers
| C22H46O | |
| MFCD00002939 | |
| 1-docosanol, behenyl alcohol, docosanol, behenic alcohol, docosyl alcohol, n-docosanol, abreva, tadenan, lidavol, lanette 22 | |
| CHEBI:31000 | |
| CCCCCCCCCCCCCCCCCCCCCCO |
Specifications
| 661-19-8 | |
| White | |
| MFCD00002939 | |
| NOPFSRXAKWQILS-UHFFFAOYSA-N | |
| docosan-1-ol | |
| 12620 | |
| 326.61 | |
| Pellets |
| 72°C | |
| C22H46O | |
| 1-docosanol, behenyl alcohol, docosanol, behenic alcohol, docosyl alcohol, n-docosanol, abreva, tadenan, lidavol, lanette 22 | |
| CCCCCCCCCCCCCCCCCCCCCCO | |
| 326.609 | |
| CHEBI:31000 | |
| ≥98.0% (GC) | |
| 1-Docosanol |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : JR1315000
TSCA : Yes