accessibility menu, dialog, popup

UserName

CAS RN 661-19-8

IUPAC Name: docosan-1-ol
Synonyms: docosanol abreva lidavol tadenan stenol 1822 stenol 1822A lanette 22
Molecular Weight (g/mol): 326.61
Molecular Formula: C22H46O
InChi Key: NOPFSRXAKWQILS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCO
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)