missing translation for 'onlineSavingsMsg'
Learn More

1-Docosanol 98.0+%, TCI America™

Catalog No. D096425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D096425G 25 g
D0964500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D096425G Supplier TCI America Supplier No. D096425G

Chemical Identifiers

CAS 661-19-8
Molecular Formula C22H46O
Molecular Weight (g/mol) 326.609
MDL Number MFCD00002939
InChI Key NOPFSRXAKWQILS-UHFFFAOYSA-N
Synonym 1-docosanol, behenyl alcohol, docosanol, behenic alcohol, docosyl alcohol, n-docosanol, abreva, tadenan, lidavol, lanette 22
PubChem CID 12620
ChEBI CHEBI:31000
IUPAC Name docosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 72°C
Color White
Quantity 25 g
Formula Weight 326.61
Percent Purity ≥98.0% (GC)
Physical Form Pellets
Chemical Name or Material 1-Docosanol
EINECSNumber (2)-0217
RTECSNumber JR1315000
TSCA Yes