missing translation for 'onlineSavingsMsg'
Learn More

1-Difluoromethoxy-3-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog No. p-7026321
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2226714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2226714 Supplier Thermo Scientific Chemicals Supplier No. B2226714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22236-07-3 | Molecular Formula: C7H5F2NO3 | Molecular Weight: 189.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22236-07-3
Molecular Formula C7H5F2NO3
Molecular Weight (g/mol) 189.12
MDL Number MFCD03407974
InChI Key NYVCZALWNPMMSQ-UHFFFAOYSA-N
Synonym 1-difluoromethoxy-3-nitrobenzene, 3-difluoromethoxy nitrobenzene, benzene, 1-difluoromethoxy-3-nitro, acmc-1coex, nyvczalwnpmmsq-uhfffaoysa, difluoromethyl 3-nitrophenyl ether, 3-difluoromethoxy-1-nitrobenzene, 3-nitro-1-difluoromethoxy benzene, 3-nitro-alpha,alpha-difluoroanisole, alpha,alpha-difluoro-3-nitroanisole
PubChem CID 2774117
IUPAC Name 1-(difluoromethoxy)-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(OC(F)F)=C1

Specifications

Density 1.391
Boiling Point 240°C to 242°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 25 g
Formula Weight 189.12
Percent Purity 98%
Chemical Name or Material 1-Difluoromethoxy-3-nitrobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only