Learn More
1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
CAS: 1151802-22-0 | C12H19BN2O2 | 234.106 g/mol
Supplier: Thermo Scientific Chemicals H6667203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester | |
| C12H19BN2O2 | |
| 1 g | |
| 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopropyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopropyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-di..., 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopropyl-4-pinacolylboronate-pyrazole, 1-cyclopropy-1-yl-boronic acid pinacol ester, 1-cyclopropyl 1h-pyrazole-4-boronic acid pinacol ester, 1h-pyrazole,1-cyclopropyl-4-4,4,5,5-tetramethyl-1, 1-cyclopropyl-1h-pyrazol-4-yl boronic acid pinacol ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CC3 | |
| 234.106 | |
| 234.11 | |
| Solid |
| 1151802-22-0 | |
| MFCD16659007 | |
| 19218005 | |
| NLWYVKHISUTBMY-UHFFFAOYSA-N | |
| 1-cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole | |
| 59327133 | |
| 97% |
Chemical Identifiers
| 1151802-22-0 | |
| 234.106 | |
| NLWYVKHISUTBMY-UHFFFAOYSA-N | |
| 59327133 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CC3 |
| C12H19BN2O2 | |
| MFCD16659007 | |
| 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopropyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopropyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-di..., 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopropyl-4-pinacolylboronate-pyrazole, 1-cyclopropy-1-yl-boronic acid pinacol ester, 1-cyclopropyl 1h-pyrazole-4-boronic acid pinacol ester, 1h-pyrazole,1-cyclopropyl-4-4,4,5,5-tetramethyl-1, 1-cyclopropyl-1h-pyrazol-4-yl boronic acid pinacol ester | |
| 1-cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only