missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%

Catalog No. p-7057223
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66672MD 250 mg
AAH6667203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66672MD Supplier Thermo Scientific Chemicals Supplier No. H66672MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1151802-22-0 | C12H19BN2O2 | 234.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1151802-22-0
Molecular Formula C12H19BN2O2
Molecular Weight (g/mol) 234.106
MDL Number MFCD16659007
InChI Key NLWYVKHISUTBMY-UHFFFAOYSA-N
Synonym 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopropyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopropyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-di..., 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopropyl-4-pinacolylboronate-pyrazole, 1-cyclopropy-1-yl-boronic acid pinacol ester, 1-cyclopropyl 1h-pyrazole-4-boronic acid pinacol ester, 1h-pyrazole,1-cyclopropyl-4-4,4,5,5-tetramethyl-1, 1-cyclopropyl-1h-pyrazol-4-yl boronic acid pinacol ester
PubChem CID 59327133
IUPAC Name 1-cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CC3

Specifications

Quantity 250 mg
Beilstein 19218005
Formula Weight 234.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only