accessibility menu, dialog, popup

UserName

CAS RN 1151802-22-0

IUPAC Name: 1-cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 234.11
Molecular Formula: C12H19BN2O2
InChi Key: NLWYVKHISUTBMY-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN(N=C1)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results