Learn More
1-Chloronaphthalene, tech. 85%, remainder 2-chloronaphthalene
CAS: 90-13-1 | C10H7Cl | 162.616 g/mol
Supplier: Thermo Scientific Chemicals A150290B
Description
1-Chloronaphthalene was used as solvent additive in the fabrication of fullerene based photovoltaic devices. It was used in preparation of polyvinyl chloride based membranes of 5,10,15,20-tetrakis(4-methoxyphenyl)porphyrinatocobalt(II) for arsenite determination. Also used in the synthesis of pharmaceutical compounds such as aryl chlorides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chloronaphthalene was used as solvent additive in the fabrication of fullerene based photovoltaic devices. It was used in preparation of polyvinyl chloride based membranes of 5,10,15,20-tetrakis(4-methoxyphenyl)porphyrinatocobalt(II) for arsenite determination. Also used in the synthesis of pharmaceutical compounds such as aryl chlorides.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 1-Chloronaphthalene | |
| ∼-8°C | |
| 259°C | |
| remainder 2-chloronaphthalene | |
| 1.633 | |
| 1000 g | |
| 14,2149 | |
| Insoluble in water. | |
| C1=CC=C2C(=C1)C=CC=C2Cl | |
| 162.616 | |
| 162.62 | |
| Technical |
| 90-13-1 | |
| 1.194 | |
| 121°C (249°F) | |
| C10H7Cl | |
| MFCD00003874 | |
| 970836 | |
| alpha-chloronaphthalene, naphthalene, 1-chloro, 1-naphthyl chloride, chloronaphthalene, 1-chloro-naphthalene, alpha-naphthyl chloride, 1-chlornaftalen, 1-chloro naphthalene, alpha-chlornaphthalene, .alpha.-chloronaphthalene | |
| JTPNRXUCIXHOKM-UHFFFAOYSA-N | |
| 1-chloronaphthalene | |
| 7003 | |
| 85% |
Chemical Identifiers
| 90-13-1 | |
| 162.616 | |
| JTPNRXUCIXHOKM-UHFFFAOYSA-N | |
| 7003 | |
| C1=CC=C2C(=C1)C=CC=C2Cl |
| C10H7Cl | |
| MFCD00003874 | |
| alpha-chloronaphthalene, naphthalene, 1-chloro, 1-naphthyl chloride, chloronaphthalene, 1-chloro-naphthalene, alpha-naphthyl chloride, 1-chlornaftalen, 1-chloro naphthalene, alpha-chlornaphthalene, .alpha.-chloronaphthalene | |
| 1-chloronaphthalene |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 201-967-3
RTECSNumber : QJ2100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only