accessibility menu, dialog, popup

UserName

Cl

CAS RN 90-13-1

IUPAC Name: 1-chloronaphthalene
Synonyms: 1-chloronaphthalene
Molecular Weight (g/mol): 162.62
Molecular Formula: C10H7Cl
InChi Key: JTPNRXUCIXHOKM-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (5)
Molecular Weight (g/mol) 
  • (8)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (5)
  • (6)
  • (3)
  • (3)
  • (5)
  • (8)
  • (3)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (8)
  • (6)