missing translation for 'onlineSavingsMsg'
Learn More

1-Chloronaphthalene 97.0+%, TCI America™

Catalog No. p-7125400
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C23105G 5 g
C231025G 25 g
2 options

Catalog No. C23105G

Supplier: TCI America C23105G

Only null left

Chemical Identifiers

CAS 90-13-1
Molecular Formula C10H7Cl
Molecular Weight (g/mol) 162.616
MDL Number MFCD00003874
InChI Key JTPNRXUCIXHOKM-UHFFFAOYSA-N
Synonym alpha-chloronaphthalene, naphthalene, 1-chloro, 1-naphthyl chloride, chloronaphthalene, 1-chloro-naphthalene, alpha-naphthyl chloride, 1-chlornaftalen, 1-chloro naphthalene, alpha-chlornaphthalene, .alpha.-chloronaphthalene
PubChem CID 7003
IUPAC Name 1-chloronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Cl

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 263°C
Quantity 5 g
UN Number 3082
Formula Weight 162.62
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloronaphthalene
EINECSNumber (4)-0316
RTECSNumber QJ2100000
TSCA Yes