missing translation for 'onlineSavingsMsg'
Learn More

1-Chloronaphthalene 85.0+%, TCI America™

Catalog No. p-7124284
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C021225G 25 g
C0212100G 100 g
C0212500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C021225G Supplier TCI America Supplier No. C021225G
Only null left

Chemical Identifiers

CAS 90-13-1
Molecular Formula C10H7Cl
Molecular Weight (g/mol) 162.616
MDL Number MFCD00003874
InChI Key JTPNRXUCIXHOKM-UHFFFAOYSA-N
Synonym alpha-chloronaphthalene, naphthalene, 1-chloro, 1-naphthyl chloride, chloronaphthalene, 1-chloro-naphthalene, alpha-naphthyl chloride, 1-chlornaftalen, 1-chloro naphthalene, alpha-chlornaphthalene, .alpha.-chloronaphthalene
PubChem CID 7003
IUPAC Name 1-chloronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Cl

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 263°C
Quantity 25 g
UN Number 3082
Formula Weight 162.62
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloronaphthalene
EINECSNumber (4)-0316
RTECSNumber QJ2100000
TSCA Yes