missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorohexane 95.0+%, TCI America™
$46.60 - $266.56
Chemical Identifiers
| CAS | 544-10-5 |
|---|---|
| Molecular Formula | C6H13Cl |
| Molecular Weight (g/mol) | 120.62 |
| MDL Number | MFCD00001018 |
| InChI Key | MLRVZFYXUZQSRU-UHFFFAOYSA-N |
| Synonym | chlorohexane, hexyl chloride, hexane, 1-chloro, n-hexyl chloride, 1-chloro-hexane, hexane, chloro, unii-r5l7i6o9nw, r5l7i6o9nw, triisobutenyl chloride, hexylchloride |
| PubChem CID | 10992 |
| IUPAC Name | 1-chlorohexane |
| SMILES | CCCCCCCl |
Chemical Identifiers
| 544-10-5 | |
| 120.62 | |
| MLRVZFYXUZQSRU-UHFFFAOYSA-N | |
| 10992 | |
| CCCCCCCl |
| C6H13Cl | |
| MFCD00001018 | |
| chlorohexane, hexyl chloride, hexane, 1-chloro, n-hexyl chloride, 1-chloro-hexane, hexane, chloro, unii-r5l7i6o9nw, r5l7i6o9nw, triisobutenyl chloride, hexylchloride | |
| 1-chlorohexane |
Specifications
| 544-10-5 | |
| 135°C | |
| MFCD00001018 | |
| 1993 | |
| MLRVZFYXUZQSRU-UHFFFAOYSA-N | |
| 1-chlorohexane | |
| 10992 | |
| ≥95.0% (GC) | |
| 1-Chlorohexane |
| Yellow | |
| C6H13Cl | |
| 25 mL | |
| chlorohexane, hexyl chloride, hexane, 1-chloro, n-hexyl chloride, 1-chloro-hexane, hexane, chloro, unii-r5l7i6o9nw, r5l7i6o9nw, triisobutenyl chloride, hexylchloride | |
| CCCCCCCl | |
| 120.62 | |
| 120.62 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0066
TSCA : Yes