missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorohexane 95.0+%, TCI America™

Supplier:  TCI America C0184500ML

 View more versions of this product

Catalog No. C0184500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Chlorohexane
Yellow
C6H13Cl
500 mL
chlorohexane, hexyl chloride, hexane, 1-chloro, n-hexyl chloride, 1-chloro-hexane, hexane, chloro, unii-r5l7i6o9nw, r5l7i6o9nw, triisobutenyl chloride, hexylchloride
CCCCCCCl
120.62
120.62
Liquid
544-10-5
135°C
MFCD00001018
1993
MLRVZFYXUZQSRU-UHFFFAOYSA-N
1-chlorohexane
10992
≥95.0% (GC)

Chemical Identifiers

544-10-5
120.62
MLRVZFYXUZQSRU-UHFFFAOYSA-N
10992
CCCCCCCl
C6H13Cl
MFCD00001018
chlorohexane, hexyl chloride, hexane, 1-chloro, n-hexyl chloride, 1-chloro-hexane, hexane, chloro, unii-r5l7i6o9nw, r5l7i6o9nw, triisobutenyl chloride, hexylchloride
1-chlorohexane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0066

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions