missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloroethyl Isopropyl Carbonate 98.0+%, TCI America™
Supplier: TCI America C1787250G
Specifications
| 1-Chloroethyl Isopropyl Carbonate | |
| Colorless | |
| C6H11ClO3 | |
| 250 g | |
| Carbonic Acid 1-Chloroethyl Isopropyl Ester | |
| CC(C)OC(=O)OC(C)Cl | |
| 166.601 | |
| 166.60 | |
| Liquid |
| 98298-66-9 | |
| 70°C | |
| MFCD04038100 | |
| 2920 | |
| XPTPAIJDVFQPJT-UHFFFAOYSA-N | |
| 1-chloroethyl propan-2-yl carbonate | |
| 11789585 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 98298-66-9 | |
| 166.601 | |
| XPTPAIJDVFQPJT-UHFFFAOYSA-N | |
| 11789585 | |
| CC(C)OC(=O)OC(C)Cl |
| C6H11ClO3 | |
| MFCD04038100 | |
| Carbonic Acid 1-Chloroethyl Isopropyl Ester | |
| 1-chloroethyl propan-2-yl carbonate |
Safety and Handling
TSCA : No