accessibility menu, dialog, popup

UserName

CAS RN 98298-66-9

IUPAC Name: 1-chloroethyl propan-2-yl carbonate
Synonyms: 1-chloroethyl isopropyl carbonate
Molecular Weight (g/mol): 166.6
Molecular Formula: C6H11ClO3
InChi Key: XPTPAIJDVFQPJT-UHFFFAOYNA-N
SMILES: CC(C)OC(=O)OC(C)Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results