missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroethyl Isopropyl Carbonate 98.0+%, TCI America™

Catalog No. p-7125168
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C178725G 25 g
C1787250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C178725G Supplier TCI America Supplier No. C178725G

Chemical Identifiers

CAS 98298-66-9
Molecular Formula C6H11ClO3
Molecular Weight (g/mol) 166.601
MDL Number MFCD04038100
InChI Key XPTPAIJDVFQPJT-UHFFFAOYSA-N
Synonym Carbonic Acid 1-Chloroethyl Isopropyl Ester
PubChem CID 11789585
IUPAC Name 1-chloroethyl propan-2-yl carbonate
SMILES CC(C)OC(=O)OC(C)Cl

Specifications

Color Colorless
Boiling Point 70°C
Quantity 25 g
UN Number 2920
Formula Weight 166.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloroethyl Isopropyl Carbonate