missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-fluorobenzene, 98%

Catalog No. AAA1369014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1369014 25 g
AAA1369022 100 g
AAA1369036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1369014 Supplier Thermo Scientific Chemicals Supplier No. A1369014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 352-33-0 | C6H4ClF | 130.55 g/mol

1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4'-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 '-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4′-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 ′-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 352-33-0
Molecular Formula C6H4ClF
Molecular Weight (g/mol) 130.55
MDL Number MFCD00000603
InChI Key RJCGZNCCVKIBHO-UHFFFAOYSA-N
Synonym p-chlorofluorobenzene, p-fluorochlorobenzene, 4-chlorofluorobenzene, benzene, 1-chloro-4-fluoro, 1-fluoro-4-chlorobenzene, 1-chloro-4-fluoro-benzene, 4-fluorochlorobenzene, 1,4-fluorochlorobenzene, p-chloro-fluorobenzene, pubchem3420
PubChem CID 9604
IUPAC Name 1-chloro-4-fluorobenzene
SMILES FC1=CC=C(Cl)C=C1

Specifications

Melting Point -22°C
Density 1.228
Boiling Point 129°C to 130°C
Flash Point 29°C (84°F)
Refractive Index 1.495
Quantity 25 g
UN Number UN1993
Beilstein 1904542
Solubility Information Difficult to mix.
Formula Weight 130.55
Percent Purity 98%
Chemical Name or Material 1-Chloro-4-fluorobenzene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-521-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only