accessibility menu, dialog, popup

UserName

FCl

CAS RN 352-33-0

IUPAC Name: 1-chloro-4-fluorobenzene
Synonyms: P-chlorofluorobenzene
Molecular Weight (g/mol): 130.55
Molecular Formula: C6H4ClF
InChi Key: RJCGZNCCVKIBHO-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C=C1
Boiling Point 
  • (5)
  • (1)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)