missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-4-ethynylbenzene 98.0+%, TCI America™
Supplier: TCI America C26705G
Specifications
| 1-Chloro-4-ethynylbenzene | |
| 873-73-4 | |
| 5 g | |
| C8H5Cl | |
| 1325 | |
| LFZJRTMTKGYJRS-UHFFFAOYSA-N | |
| 1-chloro-4-ethynylbenzene | |
| 70118 | |
| ≥98.0% (GC) |
| 47°C | |
| 60°C | |
| Crystal-Powder at 20°C | |
| MFCD00191917 | |
| 4-chlorophenylacetylene, benzene, 1-chloro-4-ethynyl, 4'-chlorophenyl acetylene, 4-chlorophenyl acetylene, 1-chloro-4-ethynyl-benzene, p-ethynylchlorobenzene, p-chlorophenylacetylene, p-chlorophenyl acetylene, 1-chlor-4-ethinylbenzol, 4-chlorophenylethyne | |
| C#CC1=CC=C(C=C1)Cl | |
| 136.578 | |
| 136.58 |
Chemical Identifiers
| 873-73-4 | |
| 136.578 | |
| LFZJRTMTKGYJRS-UHFFFAOYSA-N | |
| 70118 | |
| C#CC1=CC=C(C=C1)Cl |
| C8H5Cl | |
| MFCD00191917 | |
| 4-chlorophenylacetylene, benzene, 1-chloro-4-ethynyl, 4'-chlorophenyl acetylene, 4-chlorophenyl acetylene, 1-chloro-4-ethynyl-benzene, p-ethynylchlorobenzene, p-chlorophenylacetylene, p-chlorophenyl acetylene, 1-chlor-4-ethinylbenzol, 4-chlorophenylethyne | |
| 1-chloro-4-ethynylbenzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator