accessibility menu, dialog, popup

UserName

CAS RN 873-73-4

IUPAC Name: 1-chloro-4-ethynylbenzene
Synonyms: 1-chloro-4-ethynylbenzene
Molecular Weight (g/mol): 136.58
Molecular Formula: C8H5Cl
InChi Key: LFZJRTMTKGYJRS-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C#C
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)