missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-ethynylbenzene 98.0+%, TCI America™

Catalog No. p-7125591
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26701G 1 g
C26705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26701G Supplier TCI America Supplier No. C26701G
Only null left

Chemical Identifiers

CAS 873-73-4
Molecular Formula C8H5Cl
Molecular Weight (g/mol) 136.578
MDL Number MFCD00191917
InChI Key LFZJRTMTKGYJRS-UHFFFAOYSA-N
Synonym 4-chlorophenylacetylene, benzene, 1-chloro-4-ethynyl, 4'-chlorophenyl acetylene, 4-chlorophenyl acetylene, 1-chloro-4-ethynyl-benzene, p-ethynylchlorobenzene, p-chlorophenylacetylene, p-chlorophenyl acetylene, 1-chlor-4-ethinylbenzol, 4-chlorophenylethyne
PubChem CID 70118
IUPAC Name 1-chloro-4-ethynylbenzene
SMILES C#CC1=CC=C(C=C1)Cl

Specifications

Melting Point 47°C
Boiling Point 60°C
Quantity 1 g
Physical Form Crystal-Powder at 20°C
UN Number 1325
Formula Weight 136.58
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1-Chloro-4-ethynylbenzene
TSCA No
Recommended Storage Refrigerator