missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-hydroxypropane, 98%, stabilized

CAS: 627-30-5 | C3H7ClO | 94.54 g/mol

Supplier:  Thermo Scientific Chemicals 109291000

 View more versions of this product

Catalog No. AC109291000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Chloro-3-hydroxypropane
627-30-5
100.0
Colorless to Yellow
160°C to 162°C
Authentic
Glass bottle
1.4440 to 1.446
MFCD00002943
01, 356
3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1
LAMUXTNQCICZQX-UHFFFAOYSA-N
3-chloropropan-1-ol
0.1% Na2CO3
6 mPa.s (20°C)
98%
Stabilized
97.5
-20°C
1.1300g/mL
75°C
97.5% min. (GC)
C3H7ClO
Cl(CH2)3OH
100 g
1.13
Solubility in water: 300g/L water (20°C). Other solubilities: reacts with water forming hcl
C(CO)CCl
94.54
12313
94.54
Liquid

Chemical Identifiers

627-30-5
94.54
LAMUXTNQCICZQX-UHFFFAOYSA-N
12313
C(CO)CCl
C3H7ClO
MFCD00002943
3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1
3-chloropropan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

EINECSNumber : 210-992-9

Recommended Storage : Normal conditions

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only