missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-hydroxypropane, 98%, stabilized

Catalog No. p-3757231
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109290250 25 g Glass bottle
AC109291000 100 g Glass bottle
AC109295000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC109290250 Supplier Thermo Scientific Chemicals Supplier No. 109290250
Only null left

CAS: 627-30-5 | C3H7ClO | 94.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 627-30-5
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.54
MDL Number MFCD00002943
InChI Key LAMUXTNQCICZQX-UHFFFAOYSA-N
Synonym 3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1
PubChem CID 12313
IUPAC Name 3-chloropropan-1-ol
SMILES C(CO)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -20°C
Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 160°C to 162°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4440 to 1.446
Linear Formula Cl(CH2)3OH
Quantity 25 g
Beilstein 01, 356
Specific Gravity 1.13
Solubility Information Solubility in water: 300g/L water (20°C). Other solubilities: reacts with water forming hcl
Stabilizer 0.1% Na2CO3
Viscosity 6 mPa.s (20°C)
Formula Weight 94.54
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Chloro-3-hydroxypropane, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 210-992-9
Recommended Storage Normal conditions

RUO – Research Use Only