accessibility menu, dialog, popup

UserName

CAS RN 627-30-5

IUPAC Name: 3-chloropropan-1-ol
Synonyms: 3-chloro-1-propanol
Molecular Weight (g/mol): 94.54
Molecular Formula: C3H7ClO
InChi Key: LAMUXTNQCICZQX-UHFFFAOYSA-N
SMILES: OCCCCl
Boiling Point 
  • (3)
  • (6)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)