missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-2-naphthol 98.0+%, TCI America™
Supplier: TCI America C31751G
Specifications
| 1-Chloro-2-naphthol | |
| 68°C | |
| C10H7ClO | |
| 1 g | |
| C1=CC=C2C(=C1)C=CC(=C2Cl)O | |
| 178.615 | |
| 178.62 | |
| Crystalline Powder |
| 633-99-8 | |
| 157°C | |
| MFCD00997084 | |
| RMSOEGBYNWXXBG-UHFFFAOYSA-N | |
| 1-chloronaphthalen-2-ol | |
| 12459 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 633-99-8 | |
| 178.615 | |
| RMSOEGBYNWXXBG-UHFFFAOYSA-N | |
| 1-chloronaphthalen-2-ol |
| C10H7ClO | |
| MFCD00997084 | |
| 12459 | |
| C1=CC=C2C(=C1)C=CC(=C2Cl)O |
Safety and Handling
RTECSNumber : QL3410000
TSCA : No