CAS RN 633-99-8
CAS RN 633-99-8
IUPAC Name:
1-chloronaphthalen-2-ol
Synonyms:
2-naphthol, 1-chloro-
Molecular Weight (g/mol):
178.62
Molecular Formula:
C10H7ClO
InChi Key:
RMSOEGBYNWXXBG-UHFFFAOYSA-N
SMILES:
OC1=CC=C2C=CC=CC2=C1Cl
1
–
1
of
1
results
1
1-Chloro-2-naphthol 98.0+%, TCI America™
CAS: 633-99-8 Molecular Formula: C10H7ClO Molecular Weight (g/mol): 178.615 MDL Number: MFCD00997084 InChI Key: RMSOEGBYNWXXBG-UHFFFAOYSA-N PubChem CID: 12459 IUPAC Name: 1-chloronaphthalen-2-ol SMILES: C1=CC=C2C(=C1)C=CC(=C2Cl)O