missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,4-dinitrobenzene 99.0+%, TCI America™

Supplier:  TCI America C016225G

 View more versions of this product

Catalog No. C016225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Chloro-2,4-dinitrobenzene
52°C
C6H3ClN2O4
25 g
2,4-dinitrochlorobenzene, dinitrochlorobenzene, dncb, cdnb, chlorodinitrobenzene, 4-chloro-1,3-dinitrobenzene, 2,4-dinitrophenyl chloride, benzene, 1-chloro-2,4-dinitro, dinitrochlorobenzol, 1,3-dinitro-4-chlorobenzene
[O-][N+](=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
202.55
CHEBI:34718
≥99.0% (GC)
97-00-7
315°C
MFCD00007075
3441
VYZAHLCBVHPDDF-UHFFFAOYSA-N
1-chloro-2,4-dinitrobenzene
6
202.55
Crystalline Powder

Chemical Identifiers

97-00-7
202.55
VYZAHLCBVHPDDF-UHFFFAOYSA-N
6
1-chloro-2,4-dinitrobenzene
C6H3ClN2O4
MFCD00007075
2,4-dinitrochlorobenzene, dinitrochlorobenzene, dncb, cdnb, chlorodinitrobenzene, 4-chloro-1,3-dinitrobenzene, 2,4-dinitrophenyl chloride, benzene, 1-chloro-2,4-dinitro, dinitrochlorobenzol, 1,3-dinitro-4-chlorobenzene
CHEBI:34718
[O-][N+](=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0454

RTECSNumber : CZ0525000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions