missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,4-dinitrobenzene 99.0+%, TCI America™

Catalog No. p-7124253
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C016225G 25 g
C0162500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. C016225G missing translation for 'mfr' TCI America missing translation for 'supplierNo' C016225G
Only null left

Chemical Identifiers

CAS 97-00-7
Molecular Formula C6H3ClN2O4
Molecular Weight (g/mol) 202.55
MDL Number MFCD00007075
InChI Key VYZAHLCBVHPDDF-UHFFFAOYSA-N
Synonym 2,4-dinitrochlorobenzene, dinitrochlorobenzene, dncb, cdnb, chlorodinitrobenzene, 4-chloro-1,3-dinitrobenzene, 2,4-dinitrophenyl chloride, benzene, 1-chloro-2,4-dinitro, dinitrochlorobenzol, 1,3-dinitro-4-chlorobenzene
PubChem CID 6
ChEBI CHEBI:34718
IUPAC Name 1-chloro-2,4-dinitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 52°C
Boiling Point 315°C
Quantity 25 g
UN Number 3441
Formula Weight 202.55
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloro-2,4-dinitrobenzene
EINECSNumber (3)-0454
RTECSNumber CZ0525000
TSCA Yes