missing translation for 'onlineSavingsMsg'
Learn More

1-Butylthiourea 98.0+%, TCI America™

Catalog No. p-7123875
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49711G 1 g
B49715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49711G Supplier TCI America Supplier No. B49711G
Only null left

Chemical Identifiers

CAS 1516-32-1
Molecular Formula C5H12N2S
Molecular Weight (g/mol) 132.23
MDL Number MFCD00022173
InChI Key GMEGXJPUFRVCPX-UHFFFAOYSA-N
Synonym 1-butyl-2-thiourea, n-butylthiourea, thiourea, butyl, n-butyl thiourea, butyl-2-thiourea, thiourea, n-butyl, usaf d-5, 1-butylthiourea, urea, 1-butyl-2-thio, n-n-butyl thiourea
PubChem CID 1551919
IUPAC Name butylthiourea
SMILES CCCCNC(N)=S

Specifications

Melting Point 77°C
Color White
Quantity 1 g
Formula Weight 132.23
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butylthiourea
RTECSNumber YS4375000
TSCA Yes