missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-4-chlorobenzene 97.0+%, TCI America™
Supplier: TCI America B314325G
Specifications
| 1-Butyl-4-chlorobenzene | |
| Yellow | |
| 25 g | |
| C10H13Cl | |
| 4-butylchlorobenzene, 4-n-butylchlorobenzene, 1-n-butyl-4-chlorobenzene, p-clorobutylbenzene, 4-chlorophenylbutane, acmc-209dbt, 4-n-butyl-chlorobenzene, 4-butyl-1-chlorobenzene, 1-chloro-4-butylbenzene, benzene, 1-butyl-4-chloro | |
| CCCCC1=CC=C(C=C1)Cl | |
| 168.664 | |
| 168.66 |
| 15499-27-1 | |
| 228°C | |
| Clear Liquid at 20°C | |
| MFCD00051926 | |
| SKNUPXIXICTRJE-UHFFFAOYSA-N | |
| 1-butyl-4-chlorobenzene | |
| 524314 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 15499-27-1 | |
| 168.664 | |
| SKNUPXIXICTRJE-UHFFFAOYSA-N | |
| 524314 | |
| CCCCC1=CC=C(C=C1)Cl |
| C10H13Cl | |
| MFCD00051926 | |
| 4-butylchlorobenzene, 4-n-butylchlorobenzene, 1-n-butyl-4-chlorobenzene, p-clorobutylbenzene, 4-chlorophenylbutane, acmc-209dbt, 4-n-butyl-chlorobenzene, 4-butyl-1-chlorobenzene, 1-chloro-4-butylbenzene, benzene, 1-butyl-4-chloro | |
| 1-butyl-4-chlorobenzene |
Safety and Handling
TSCA : No