missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-4-chlorobenzene 97.0+%, TCI America™

Catalog No. p-7119981
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31435G 5 g
B314325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31435G Supplier TCI America Supplier No. B31435G

Chemical Identifiers

CAS 15499-27-1
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.664
MDL Number MFCD00051926
InChI Key SKNUPXIXICTRJE-UHFFFAOYSA-N
Synonym 4-butylchlorobenzene, 4-n-butylchlorobenzene, 1-n-butyl-4-chlorobenzene, p-clorobutylbenzene, 4-chlorophenylbutane, acmc-209dbt, 4-n-butyl-chlorobenzene, 4-butyl-1-chlorobenzene, 1-chloro-4-butylbenzene, benzene, 1-butyl-4-chloro
PubChem CID 524314
IUPAC Name 1-butyl-4-chlorobenzene
SMILES CCCCC1=CC=C(C=C1)Cl

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 228°C
Formula Weight 168.66
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1-Butyl-4-chlorobenzene