CAS RN 15499-27-1
IUPAC Name:
1-butyl-4-chlorobenzene
Synonyms:
1-butyl-4-chlorobenzene
Molecular Weight (g/mol):
168.66
Molecular Formula:
C10H13Cl
InChi Key:
SKNUPXIXICTRJE-UHFFFAOYSA-N
SMILES:
CCCCC1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
1-Butyl-4-chlorobenzene 97.0+%, TCI America™
CAS: 15499-27-1 Molecular Formula: C10H13Cl Molecular Weight (g/mol): 168.664 MDL Number: MFCD00051926 InChI Key: SKNUPXIXICTRJE-UHFFFAOYSA-N Synonym: 4-butylchlorobenzene,4-n-butylchlorobenzene,1-n-butyl-4-chlorobenzene,p-clorobutylbenzene,4-chlorophenylbutane,acmc-209dbt,4-n-butyl-chlorobenzene,4-butyl-1-chlorobenzene,1-chloro-4-butylbenzene,benzene, 1-butyl-4-chloro PubChem CID: 524314 IUPAC Name: 1-butyl-4-chlorobenzene SMILES: CCCCC1=CC=C(C=C1)Cl