missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-2-nitrobenzene 96.0+%, TCI America™
Supplier: TCI America B309125G
Specifications
| 1-Butyl-2-nitrobenzene | |
| Yellow | |
| C10H13NO2 | |
| 25 g | |
| CCCCC1=CC=CC=C1[N+]([O-])=O | |
| 179.22 | |
| 179.22 | |
| Liquid |
| 7137-55-5 | |
| 127°C | |
| MFCD00130028 | |
| RSVIEXUVWMYRGB-UHFFFAOYSA-N | |
| 1-butyl-2-nitrobenzene | |
| 81555 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 7137-55-5 | |
| 179.22 | |
| RSVIEXUVWMYRGB-UHFFFAOYSA-N | |
| 1-butyl-2-nitrobenzene |
| C10H13NO2 | |
| MFCD00130028 | |
| 81555 | |
| CCCCC1=CC=CC=C1[N+]([O-])=O |
Safety and Handling
TSCA : No