accessibility menu, dialog, popup

UserName

H3CNOO

CAS RN 7137-55-5

IUPAC Name: 1-butyl-2-nitrobenzene
Synonyms: 1-butyl-2-nitrobenzene
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: RSVIEXUVWMYRGB-UHFFFAOYSA-N
SMILES: CCCCC1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results