missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-2,3-dimethylimidazolium Trifluoromethanesulfonate 98.0+%, TCI America™
Supplier: TCI America B418225G
Specifications
| 1-Butyl-2,3-dimethylimidazolium Trifluoromethanesulfonate | |
| 44°C | |
| C10H17F3N2O3S | |
| 25 g | |
| KSOGGGZFEJTGPZ-UHFFFAOYSA-M | |
| 1-butyl-2,3-dimethylimidazol-3-ium;trifluoromethanesulfonate | |
| 2734250 | |
| ≥98.0% (HPLC,N) |
| 765910-73-4 | |
| White | |
| MFCD03427624 | |
| 1-butyl-2,3-dimethylimidazolium trifluoromethanesulfonate, 1-butyl-2,3-dimethylimidazolium triflate, dsstox_cid_29315, dsstox_rid_83431, dsstox_gsid_49356, ksc496s6h, 1-butyl-2,3-dimethylimidazol-1-ium triflate, 3-butyl-1,2-dimethylimidazol-1-ium triflate, 1-butyl-2,3-dimethylimidazolium trifluoromethansulfonate, 1-butyl-2,3-dimethylimidazol-3-ium; trifluoromethanesulfonate | |
| CCCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[O-] | |
| 302.312 | |
| 302.31 | |
| Crystalline Powder |
Chemical Identifiers
| 765910-73-4 | |
| 302.312 | |
| KSOGGGZFEJTGPZ-UHFFFAOYSA-M | |
| 2734250 | |
| CCCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[O-] |
| C10H17F3N2O3S | |
| MFCD03427624 | |
| 1-butyl-2,3-dimethylimidazolium trifluoromethanesulfonate, 1-butyl-2,3-dimethylimidazolium triflate, dsstox_cid_29315, dsstox_rid_83431, dsstox_gsid_49356, ksc496s6h, 1-butyl-2,3-dimethylimidazol-1-ium triflate, 3-butyl-1,2-dimethylimidazol-1-ium triflate, 1-butyl-2,3-dimethylimidazolium trifluoromethansulfonate, 1-butyl-2,3-dimethylimidazol-3-ium; trifluoromethanesulfonate | |
| 1-butyl-2,3-dimethylimidazol-3-ium;trifluoromethanesulfonate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator