missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-2,3-dimethylimidazolium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7123395
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41825G 5 g
B418225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41825G Supplier TCI America Supplier No. B41825G

Chemical Identifiers

CAS 765910-73-4
Molecular Formula C10H17F3N2O3S
Molecular Weight (g/mol) 302.312
MDL Number MFCD03427624
InChI Key KSOGGGZFEJTGPZ-UHFFFAOYSA-M
Synonym 1-butyl-2,3-dimethylimidazolium trifluoromethanesulfonate, 1-butyl-2,3-dimethylimidazolium triflate, dsstox_cid_29315, dsstox_rid_83431, dsstox_gsid_49356, ksc496s6h, 1-butyl-2,3-dimethylimidazol-1-ium triflate, 3-butyl-1,2-dimethylimidazol-1-ium triflate, 1-butyl-2,3-dimethylimidazolium trifluoromethansulfonate, 1-butyl-2,3-dimethylimidazol-3-ium; trifluoromethanesulfonate
PubChem CID 2734250
IUPAC Name 1-butyl-2,3-dimethylimidazol-3-ium;trifluoromethanesulfonate
SMILES CCCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 44°C
Color White
Quantity 5 g
Formula Weight 302.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium Trifluoromethanesulfonate
TSCA No
Recommended Storage Refrigerator