accessibility menu, dialog, popup

UserName

CAS RN 765910-73-4

IUPAC Name: 1-butyl-2,3-dimethyl-1H-imidazol-3-ium trifluoromethanesulfonate
Synonyms: 3-butyl-1,2-dimethylimidazol-1-ium triflate
Molecular Weight (g/mol): 302.31
Molecular Formula: C10H17F3N2O3S
InChi Key: KSOGGGZFEJTGPZ-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.CCCCN1C=C[N+](C)=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results