missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-1-phenylhydrazine 98.0+%, TCI America™

Catalog No. p-7119576
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B22435G 5 g
B224325G 25 g
2 options

Catalog No. B22435G

Supplier: TCI America B22435G

Only null left

Chemical Identifiers

CAS 61715-75-1
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD03844777
InChI Key VKXUWCNLFZXHEF-UHFFFAOYSA-N
PubChem CID 13042771
IUPAC Name 1-butyl-1-phenylhydrazine
SMILES CCCCN(C1=CC=CC=C1)N

Specifications

Color Yellow
Boiling Point 105°C
Quantity 5 g
Formula Weight 164.25
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Butyl-1-phenylhydrazine