accessibility menu, dialog, popup

UserName

CAS RN 61715-75-1

IUPAC Name: 1-butyl-1-phenylhydrazine
Synonyms: 1-butyl-1-phenylhydrazine
Molecular Weight (g/mol): 164.25
Molecular Formula: C10H16N2
InChi Key: VKXUWCNLFZXHEF-UHFFFAOYSA-N
SMILES: CCCCN(N)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results