missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-1-methylpiperidinium Bromide 97.0+%, TCI America™
Supplier: TCI America B342425G
Specifications
| 1-Butyl-1-methylpiperidinium Bromide | |
| 230°C | |
| C10H22BrN | |
| 25 g | |
| [Br-].CCCC[N+]1(C)CCCCC1 | |
| 236.20 | |
| 236.20 | |
| Crystalline Powder |
| 94280-72-5 | |
| White-Yellow | |
| MFCD11870112 | |
| WYABBCZMFVULEF-UHFFFAOYSA-M | |
| 1-butyl-1-methylpiperidin-1-ium bromide | |
| 19838382 | |
| ≥97.0% (T) |
Chemical Identifiers
| 94280-72-5 | |
| 236.20 | |
| WYABBCZMFVULEF-UHFFFAOYSA-M | |
| 1-butyl-1-methylpiperidin-1-ium bromide |
| C10H22BrN | |
| MFCD11870112 | |
| 19838382 | |
| [Br-].CCCC[N+]1(C)CCCCC1 |
Safety and Handling
TSCA : No