missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-1-methylpiperidinium Bromide 97.0+%, TCI America™

Catalog No. p-7122940
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34245G 5 g
B342425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34245G Supplier TCI America Supplier No. B34245G
Only null left

Chemical Identifiers

CAS 94280-72-5
Molecular Formula C10H22BrN
Molecular Weight (g/mol) 236.20
MDL Number MFCD11870112
InChI Key WYABBCZMFVULEF-UHFFFAOYSA-M
PubChem CID 19838382
IUPAC Name 1-butyl-1-methylpiperidin-1-ium bromide
SMILES [Br-].CCCC[N+]1(C)CCCCC1

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 5 g
Formula Weight 236.20
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-1-methylpiperidinium Bromide