missing translation for 'onlineSavingsMsg'
Learn More

1-Butoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene 98.0+%, TCI America™

Catalog No. B53645G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53645G 5 g
B53641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53645G Supplier TCI America Supplier No. B53645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 208709-55-1
Molecular Formula C19H28F2O
Molecular Weight (g/mol) 310.429
MDL Number MFCD20489249
InChI Key KYNDSYARZOJNCG-UHFFFAOYSA-N
PubChem CID 22184028
IUPAC Name 1-butoxy-2,3-difluoro-4-(4-propylcyclohexyl)benzene
SMILES CCCCOC1=C(C(=C(C=C1)C2CCC(CC2)CCC)F)F

Specifications

Color Colorless
Quantity 5 g
Formula Weight 310.43
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Butoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene