accessibility menu, dialog, popup

UserName

CAS RN 208709-55-1

IUPAC Name: 1-butoxy-2,3-difluoro-4-(4-propylcyclohexyl)benzene
Synonyms: 1-butoxy-2,3-difluoro-4-(4-propylcyclohexyl)benzene
Molecular Weight (g/mol): 310.43
Molecular Formula: C19H28F2O
InChi Key: KYNDSYARZOJNCG-UHFFFAOYSA-N
SMILES: CCCCOC1=C(F)C(F)=C(C=C1)C1CCC(CCC)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results